High-Tech Conversions NT56-1818 Cleanroom wipes, non-woven, lint-free, polyester/ cellulose blend, 18" x 18", 750/CS
Representative image only.
High-Tech Conversions Mfr # NT56-1818 – Item # WZ-15981-23
Lint-free, versatile, and highly effective
  • Converted in an ISO Class 6 cleanroom to maintain certification Polyester and cellulose blend is tough, absorbent, and abrasion-resistant Economical choice for general-purpose wiping
  • 2 year warranty
PRODUCT OPTIONS
£194.09GBP / Case of 750
£232.91GBP (incl VAT)
Cole-Parmer Essentials Dry Clean-Wipes; 100/Canister
£15.60GBP / Each
£18.72GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyester / Cellulose Blend
  • ColorWhite
  • ComplianceISO Class 6
  • Width (in)18
  • Length (in)18
  • Width (cm)46
  • Length (cm)46
  • DescriptionCleanroom Wipes, Non-Woven, Lint-Free, Polyester / Cellulose Blend, 18\" x 18\", 750/CS
  • Warranty2 year
Two year warranty
Key Features
  • Converted in an ISO Class 6 cleanroom to maintain certification Polyester and cellulose blend is tough, absorbent, and abrasion-resistant Economical choice for general-purpose wiping
  • 2 year warranty
More About this Item
Use these lightweight cleanroom wipes for light- to moderate-duty cleaning applications. The blend of virgin polyester and cellulose forms a strong fabric virtually free of particle generation. The ISO 6-certified fabric is also converted in a cleanroom facility minimizing exposure to pollutants.