This item is now discontinued. Please browse similar items
HF Scientific 10518 Dispenser for Total Chlorine, 100 Tests, 25 Ml
Representative image only.
HF Scientific Mfr # 10518 – Item # WZ-99560-25
Convenient no spill DPD powder dispensing
  • Precise dosing with simple push button operation
  • No direct powder or foil packet handling
SPECS & DESCRIPTION
Specifications & Description
  • Detector TypeTablet formulation for Oakton, LaMotte, Orbeco-Hellige, Palintest, Thermo Scientific Orion, Tintometer, and YSI meters
  • Test ParametersTotal Chlorine
  • Number of Tests Included100
  • DescriptionDispenser for Total Chlorine, 100 Tests, 25 mL
Key Features
  • Precise dosing with simple push button operation
  • No direct powder or foil packet handling
More About this Item
Deliver the correct volume of DPD reagent for free or total chlorine accurately using these portable dispensers. Simply invert the dispenser over the sample tube and press the button to dispense reagent. The powder dissolves quickly in the sample with no messy residue.
Use the DPD powder formulation for Hach, HF scientific, and Hanna colorimeters. Use the tablet formulation for LaMotte, Orbeco-Hellige, Palintest, Thermo Scientific Orion, and Tintometer units.
Dispensers provide enough DPD powder for 100 tests for 5-mL samples or order multi-packs of five dispensers with 200 tests each for 10-mL or 25-mL samples.