This item is now discontinued. Please browse similar items
Ashcroft 1327CM Pressure GaugeComparator, 500 spi
Representative image only.
Ashcroft Mfr # 1327CM-S-C – Item # WZ-68840-54
Field-portable pressure generation and test systems are ±0.1% accurate
  • Operating pressure to 10,000 psi
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Pressure500 psi
  • Min Pressure0 psi
  • Calibration MediumSAE 20 wight oil
  • DescriptionPressure GaugeComparator, 500 spi
  • Warranty1 Year
1 Year Warranty
Key Features
  • Operating pressure to 10,000 psi
  • 1 year warranty
More About this Item
Hydraulic two-stage pressure gauge comparator has a built-in shuttle valve that allows for rapid pressure increases at low pressures. A single carrying case holds the pump as well as the test gauge provided, a six-inch test gauge, accurate to ±0.1% full-scale. Test gauge provided has temperature compensation which maintains the ±0.1% full-scale accuracy over an operating temperature range of -25 to 125°F. Operating media is standard SAE 20 weight automative/machine oil.