Hayward TC10150STE True Union Ball Check Valve, 1-1/2" socket and NPT(F) connectors
Representative image only.
Hayward Mfr # TC10150STE – Item # WZ-01341-08
Unique square-cut elastomer seat provides seal at very low back pressures
PRODUCT OPTIONS
£121.80GBP / Each
£146.16GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))1.5
  • Outlet Port1-1/2" NPT(F) and socket
  • Connection TypeNPT(F) and Socket
  • Inlet Port1-1/2" NPT(F) and socket
  • Cv55
  • Max Temperature (° F)140
  • Max Temperature (° C)60
  • Wetted MaterialsPVC, EPDM
  • Working Pressure (PSI)150
  • DescriptionTrue Union Ball Check Valve, 1-1/2" socket and NPT(F) connectors
More About this Item
Quickly and easily install either vertically or horizontally. These valves are assembled with water soluble, silicone-free lubricant and constructed of a one-piece PVC body.