Hayward HCTN2075STV/EAU-28 electrically Actuated Ball Valve, 3-way, 3/4" CPVC
Representative image only.
Hayward Mfr # HCTN2075STV/EAU-28 – Item # WZ-01347-64
NEMA 4X polypropylene housing for harsh operating conditions
  • Thermal overload circuit protects the motor from burnout and overheating
PRODUCT OPTIONS
£503.00GBP / Each
£603.60GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.75
  • Outlet Port3/4" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port3/4" NPT(F)
  • Cv15
  • Max Pressure (PSI)150
  • Power (VAC)110
  • Max Temperature (° F)140
  • Max Temperature (° C)60
  • Wetted MaterialsCPVC, double Viton O-rings
  • Working Pressure (PSI)225
  • Actuation Motor110 VAC
  • Descriptionelectrically Actuated Ball Valve, 3-way, 3/4" CPVC
Key Features
  • Thermal overload circuit protects the motor from burnout and overheating