Hayward HCTB2075STACTV-EAU29 Electrically Actuated CPVC True Union Ball Valve, 3/4", 115 VAC
Representative image only.
Hayward Mfr # HCTB2075STACTV-EAU29 – Item # WZ-01347-22
  • Ideal for remote control of process fluids
  • Precision machined stems specifically designed for actuated valves
  • Adjustable and reversible PTFE(R) PTFE seats
  • One-piece PVC or CPVC body construction
  • Includes two Viton(R) seals for double leakproof protection
PRODUCT OPTIONS
£526.05GBP / Each
£631.26GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.75
  • Outlet Port3/4" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port3/4" NPT(F)
  • Cv15
  • Max Pressure (PSI)225
  • Power (VAC)115
  • Max Temperature (° F)194
  • Max Temperature (° C)90
  • Wetted MaterialsCPVC, Viton
  • DescriptionElectrically Actuated CPVC True Union Ball Valve, 3/4", 115 VAC
Key Features
  • Ideal for remote control of process fluids
  • Precision machined stems specifically designed for actuated valves
  • Adjustable and reversible PTFE(R) PTFE seats
  • One-piece PVC or CPVC body construction
  • Includes two Viton(R) seals for double leakproof protection
  • Remote control capabilities via an auxiliary limit switch
  • Thermal overload circuit protects the motor from burnout and overheating
  • Easy connections via a wiring terminal strip
  • Unique mounting feature allows for easy conversion to manual operation
  • Mechanical brake for precise cycle control
  • Actuator comes mounted on the valve
  • NEMA 4X polypropylene housing stands up to harsh operating conditions