Hayward FLT4202 double-length bag filter housing, 2" NPT(F)
Representative image only.
Hayward Mfr # FLV4P2S20T4VS – Item # WZ-29990-04
All-plastic construction—no need to worry about rust and corrosion!
  • Ideal for water, chemical, pharmaceutical, food and other general fluid filtration applications
  • Feature hand-removable cover and built-in basket/bag handles—simply lift out the basket and replace the filter bag
  • External cover threads are not in contact with the process media—eliminates the need for cleaning
  • Cover features buttress style threads for extra strength
  • Available in single- and double-length sizes; maximum flow rate for single-length size is 50 GPM, double-length size is 100 GPM
£2,298.03GBP / Each
£2,757.64GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Housing Material50.1
  • Port Size2" NPT(F)
  • Housing Height (in)127.25
  • Housing Outside Diameter (OD) (in)18.3
  • Housing Outside Diameter (OD) (cm)46.48
  • Descriptiondouble-length bag filter housing, 2" NPT(F)
Key Features
  • Ideal for water, chemical, pharmaceutical, food and other general fluid filtration applications
  • Feature hand-removable cover and built-in basket/bag handles—simply lift out the basket and replace the filter bag
  • External cover threads are not in contact with the process media—eliminates the need for cleaning
  • Cover features buttress style threads for extra strength
  • Available in single- and double-length sizes; maximum flow rate for single-length size is 50 GPM, double-length size is 100 GPM
  • Each filter includes an integral mounting flange and Viton(R) seal; order bags separately.