Hayward F31B37455E double-length bags; 5 µm, 50/pack
Representative image only.
Hayward Mfr # PO005G2PRSH – Item # WZ-29990-74
£382.46GBP / Pkg of 50
£458.95GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaPolypropylene, glass reinforced
  • Micron Rating5
  • Descriptiondouble-length bags; 5 µm, 50/pack
More About this Item
Hayward(R) bags are made from nonwoven polypropylene felt. The single-length bags have a surface area of 2.0 ft2; double length bags have a surface area of 4.1 ft2. Bags are available in mulitple pore sizes.