Hayward DAB1010UEE Diaphragm Valve, PVC, 1" True Union Connection
Representative image only.
Hayward Mfr # DAB1010UEE – Item # WZ-07390-51
Designed for use in corrosive environments
  • Multi-turn handle gives you precise control
  • Position indicator shows exact location of valve
  • Self draining allows for little dead volume in valve
  • PVC body and EPDM diaphragm
PRODUCT OPTIONS
£159.27GBP / Each
£191.12GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port1" true union socket
  • Connection TypeTrue union socket
  • Inlet Port1" true union socket
  • Cv13
  • Wetted MaterialsPVC
  • Working Pressure (PSI)225
  • DescriptionDiaphragm Valve, PVC, 1" True Union Connection
Key Features
  • Multi-turn handle gives you precise control
  • Position indicator shows exact location of valve
  • Self draining allows for little dead volume in valve
  • PVC body and EPDM diaphragm
More About this Item
These Hayward diaphragm valves are constructed with a PVC body and EPDM diaphragm. Valves offer a maximum temperature of 140°F (60°C) and feature true-union connections.