This item is now discontinued. Please browse similar items
Haws 8320-8325 Combination Shower with ABS plastic Eyewash
Representative image only.
Haws Mfr # 8320-8325 – Item # WZ-06767-70
Hydrodynamic shower design provides a more equal water distribution throughout the flow
  • Inverted flow sweeps contaminants away from inner eye and nasal cavity
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Eyewash Flow Rate (GPM)3.7
  • Shower Flow Rate (GPM)20
  • Bowl MaterialABS plastic
  • Shower Head MaterialABS plastic
  • Supply Pipe MaterialGalvanized steel
  • MountFloor Mount
  • Outlet Connections1-1/4\" IPS(F)
  • Inlet Connection1-1/4\" IPS(F)
  • ComplianceANSI Z358.1-2009
  • DescriptionCombination Shower with ABS plastic Eyewash
  • Warranty1 year
1 Year Warranty
Key Features
  • Inverted flow sweeps contaminants away from inner eye and nasal cavity
  • 1 year warranty
More About this Item
Combination showers feature Axion MSR 11" (27.9 cm) diameter eyewash with unique inverted flow to push contaminants away from the inner corner of the eye, consistent with first aid protocols. ABS plastic showerhead with flow control uses a hydrodynamic design to give a more equal distribution of water — provides the injured with a more thorough rinsing of the contaminant. Mesh water strainer reduces debris in the water supply for further eye protection. Constructed of galvanized steel pipe and fittings with a 9" (22.9 cm) cast iron floor flange.
Compliance: ANSI Z358.1-2009