This item is now discontinued. Please browse similar items
Harvard Apparatus 53-2802R Replacement Thumbscrews for 08024 Series Nylon/SS Clamps, 10/PK
Representative image only.
Harvard Apparatus Mfr # 53-2802R(X10 PIECES) – Item # WZ-08024-26
Glass fiber-reinforced nylon and stainless steel components provide enormous strength
  • This material has a melting point of 221°C.
  • Resistance to salt solutions and solvents.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionReplacement Thumbscrews for 08024 Series Nylon/SS Clamps, 10/PK
Key Features
  • This material has a melting point of 221°C.
  • Resistance to salt solutions and solvents.
More About this Item
Replacement thumbscrews and optional socket screws fit all frame clamps, bases, and mounting feet in rack support system. Socket screws include an Allen wrench.