This item is now discontinued. Please browse similar items
Harvard Apparatus 53-2342W Small V-Base, Nylon and SS, Supports 1 Rod Up to 1/2" Dia
Representative image only.
Harvard Apparatus Mfr # 53-2342W – Item # WZ-08024-55
Provides stability to your rack system
  • Material has a melting point of 429°F (221°C)
  • Resistance to salt solutions and solvents
SPECS & DESCRIPTION
Specifications & Description
  • Base MaterialFiberglass-reinforced nylon and stainless steel
  • Clamp TypeSmall v-base
  • Accepts Rod Diameter (in)0.5
  • DescriptionSmall V-Base, Nylon and SS, Supports 1 Rod Up to 1/2\" Dia
Key Features
  • Material has a melting point of 429°F (221°C)
  • Resistance to salt solutions and solvents
More About this Item
Bases accept rods up to 1/2'' in diameter. Neoprene feet prevent slipping; stack bases together for added stability.