This item is now discontinued. Please browse similar items
Harvard Apparatus 53-2302W Scaffold Foot, Nylon and Ss, Supports 1 Rod Up to 1/2" Dia, 4/Pk
Representative image only.
Harvard Apparatus Mfr # 53-2302W(X4 PIECES) – Item # WZ-08024-57
Provides stability to your rack system
  • Material has a melting point of 429°F (221°C)
  • Resistance to salt solutions and solvents
SPECS & DESCRIPTION
Specifications & Description
  • Base MaterialFiberglass-reinforced Nylon and Stainless Steel
  • Clamp TypeScaffold foot
  • Accepts Rod Diameter (in)0.5
  • DescriptionScaffold Foot, Nylon and Ss, Supports 1 Rod Up to 1/2\" Dia, 4/Pk
Key Features
  • Material has a melting point of 429°F (221°C)
  • Resistance to salt solutions and solvents
More About this Item
Bases accept rods up to 1/2'' in diameter. Neoprene feet prevent slipping; stack bases together for added stability.