This item is now discontinued. Please browse similar items
Harvard Apparatus 53-2232W Camera-Mount Adapter, nylon and stainless steel, 1/ea
Representative image only.
Harvard Apparatus Mfr # 53-2232W – Item # WZ-08024-51
Glass fiber-reinforced nylon and stainless steel components provide enormous strength
  • This material has a melting point of 221°C.
  • Resistance to salt solutions and solvents.
SPECS & DESCRIPTION
Specifications & Description
  • Clamp MaterialFiberglass-reinforced Nylon and Stainless Steel
  • Clamp TypeEquipment, Camera Mount
  • DescriptionCamera-Mount Adapter, nylon and stainless steel, 1/ea
Key Features
  • This material has a melting point of 221°C.
  • Resistance to salt solutions and solvents.
More About this Item
Place your equipment at odd angles - this adapter holds position anywhere in a full hemisphere. Ideal for use with T-joint clamp 08024-29.