Hamilton Microliter Low-Volume Syringe, Knurled Hub, Blunt Tip, 5 µL, 24 G

Representative image only.
Hamilton Mfr # 88000 – Item # WZ-07938-63
Designed with zero dead volume
  • Gas- and liquid-tight up to 6000 psi!
  • Needle point style 3 features a blunt needle point for use with HPLC injection valves and for sample pipetting
  • 18 month warranty

PRODUCT OPTIONS

£95.33GBP / Each
£114.40GBP (incl VAT)
Hamilton Microliter Low-Volume Syringe Repair Kit, for 5 µL Syringe
£115.15GBP / Each
£138.18GBP (incl VAT)
Often Bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Syringe TypeKnurled Hub
  • Syringe Capacity (µL)5
  • Needle Tip Style90⁰ Blunt Tip
  • Needle Tip Number3
  • Needle IncludedYes
  • Needle Gauge24
  • Needle Length (in)2.75
  • MaterialSST, tungsten
  • Outside Diameter (OD) (mm)7.82
  • DescriptionMicroliter Low-Volume Syringe, Knurled Hub, Blunt Tip, 5 µL, 24 G
  • Warranty18 months
18 Month Warranty

Key Features

  • Gas- and liquid-tight up to 6000 psi!
  • Needle point style 3 features a blunt needle point for use with HPLC injection valves and for sample pipetting
  • 18 month warranty

More About this Item

Hamilton® 7000-series microliter syringes are designed for rapid injection of minute volumes. Full syringe capacity is in the needle and is completely ejected when plunger wire is depressed—no void volume. The fine gauge plunger wire extends all the way to the tip of the needle ensuring that all solvent is expelled from the needle. Therefore, it is not recommended to remove the needle or plunger.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat