Hamilton Microliter Low-Volume Syringe, Beveled Tip, Cemented Needle, 500 µL, 22 G
Representative image only.
Hamilton Mfr # 80800 – Item # WZ-07938-05
Achieve precise liquid delivery
  • Constructed of borosilicate glass
  • Needle point style 2 features a sharp, beveled, curved, non-coring needle point recommended for piercing septa
  • Syringe features a 1000 psi max pressure and 6.81 µL dead volume
  • 18 month warranty
PRODUCT OPTIONS
£74.12GBP / Each
£88.94GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Syringe TypeCemented Needle
  • Syringe Capacity (µL)500
  • Needle IncludedYes
  • Needle Tip Number2
  • Needle Tip StyleSharp, beveled, curved, non-coring
  • Needle Gauge22
  • Needle Length (in)2
  • MaterialSST, borosilicate glass
  • Outside Diameter (OD) (mm)7.75
  • DescriptionMicroliter Low-Volume Syringe, Beveled Tip, Cemented Needle, 500 µL, 22 G
  • Warranty18 months
18 Month Warranty
Key Features
  • Constructed of borosilicate glass
  • Needle point style 2 features a sharp, beveled, curved, non-coring needle point recommended for piercing septa
  • Syringe features a 1000 psi max pressure and 6.81 µL dead volume
  • 18 month warranty
More About this Item
Hamilton® 700-series microliter syringes are designed to solve the general liquid handling requirements of manufacturing and research laboratories. Syringes are used for manual and automated GC and HPLC injections and for everyday applications that require accurate liquid measuring and dispending.
Choose from cemented needle, removable needle, or Luer tip syringes. Syringes with luer tip connections do not include a needle, order needles separately.