This item is now discontinued. Please browse similar items
Hamilton 79466 PRP-3 Reverse Phase Column, Protein Isolation, 150x4.1mm ID; 10um
Representative image only.
Hamilton Mfr # 79466 – Item # WZ-02004-10
Efficient separations with durable media
  • 18 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • PhasePolystyrenedivinylbenzene
  • Column Particle Size (µm)10
  • DescriptionPRP-3 Reverse Phase Column, Protein Isolation, 150x4.1mm ID; 10um
  • Warranty18 months
18 Month Warranty
Key Features
  • 18 month warranty
More About this Item
Hamilton polymeric PRP™ columns are resin-based, offering excellent resolution due to their high selectivity, efficiency, and capacity. The chemically inert macroporous poly(styrenedivinylbenzene)stationary phase is stable from pH 1 to 13. Compatible with any concentration of organic solvent; can be used up to 60°C and 5000 psi. Hamilton guarantees that the spherical media will not shrink or swell.
PRP-3 Reverse Phase Columns are made of a wide pore (300 Å) poly (styrene-divinylbenzene) copolymer. Use to isolate or purify proteins and peptides. For extended life, regenerate your column up to five times—protein recoveries are better than 90%. Choose a 50-mm column for the gradient elution of high-molecular-weight proteins. The 150-mm column is best suited for the gradient separation of lower molecular weight proteins (less than 5000 MW).