This item is now discontinued. Please browse similar items
Hamilton 79465 PRP-X300 Ion Exclusion Column, Analytical, 250x4.1mm ID; 7um
Representative image only.
Hamilton Mfr # 79465 – Item # WZ-01944-02
Efficient separations with durable media
  • 18 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • PhasePolystyrenedivinylbenzene
  • Column Particle Size (µm)8
  • DescriptionPRP-X300 Ion Exclusion Column, Analytical, 250x4.1mm ID; 7um
  • Warranty18 months
18 Month Warranty
Key Features
  • 18 month warranty
More About this Item
Hamilton polymeric PRP™ columns are resin-based, offering excellent resolution due to their high selectivity, efficiency, and capacity. The chemically inert macroporous poly(styrenedivinylbenzene) stationary phase is stable from pH 1 to 13. Compatible with any concentration of organic solvent; can be used up to 60°C and 5000 psi. Hamilton guarantees that the spherical media will not shrink or swell.
PRP-X300 Ion Exclusion Columns contain a partially sulfonated poly (styrene-divinylbenzene) media. Exhibit a high degree of both reverse phase and ion exclusion characteristics; separations are based on charge as well as relative hydrophobicity. Ideal for analysis of carboxylic acids and alcohols—no elevated temperature required. Exchange capacity is 0.17 mEq/g.