This item is now discontinued. Please browse similar items
Hamilton 79425 PRP-1 Reverse Phase Column, Fast Analytical, 150x4.1mm ID; 10um
Representative image only.
Hamilton Mfr # 79425 – Item # WZ-01941-02
High capacity and rapid flow rates for fast analysis
  • Compatible with any concentration of organic solvent
  • 18 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • PhasePolystyrenedivinylbenzene
  • Column Length (mm)250
  • Column Inside Diameter (ID) (mm)4.1
  • Column Particle Size (µm)10
  • Wetted MaterialsStainless steel, PSDVB/Sulfonic Acid
  • Max Pressure (PSI)5000
  • Min Operating Temperature (° C)5
  • Max Operating Temperature (° C)60
  • ApplicationsInorganic and organic cations, alkali and alkaline earth metals, mono or divalent cations
  • DescriptionPRP-1 Reverse Phase Column, Fast Analytical, 150x4.1mm ID; 10um
  • Warranty18 months
18 Month Warranty
Key Features
  • Compatible with any concentration of organic solvent
  • 18 month warranty
More About this Item
Hamilton polymeric PRP™ columns are resin-based, offering excellent resolution due to their high selectivity, efficiency, and capacity. The chemically inert macroporous poly(styrenedivinylbenzene)stationary phase is stable from pH 1 to 13. Compatible with any concentration of organic solvent; can be used up to 60°C and 5000 psi. Hamilton guarantees that the spherical media will not shrink or swell.
PRP-1 Reverse Phase Columns contain poly(styrenedivinylbenzene). Large surface area (415 m2/g) allows high capacity and rapid flow rates for fast analysis. Ideal for separating strong acids and bases. Efficiency of 30,000 to 50,000 plates/meter.