Hach TNT856 TNTplus® Spectrophotometer Chemistry, Nickel, 0.1 to 6 mg/L; 25/PK
Representative image only.
Hach Mfr # TNT856 – Item # WZ-99515-42
Perform common methods with these pre-made reagent tests
  • Measures: Nickel, 0.1 to 6 mg/L
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNickel, 0.1 to 6 mg/L
  • Compatible WithHach DR 6000, 3900, 1900
  • DescriptionSpectrophotometer Chemistry, Nickel, 0.1 to 6 mg/L; 25/PK
Key Features
  • Measures: Nickel, 0.1 to 6 mg/L
More About this Item
Dozens of options to cover most applications!