H-B Instrument B61401-5300 Liquid-in-Glass Thermometer Non-Roll Fitting, Purple PVC Plastic, Ring Top
Representative image only.
H-B Instrument Mfr # B61401-5300 – Item # WZ-08007-98
Designed to help prevent glass thermometers from rolling off a surface and breaking
  • Can be used for all types of liquid-in-glass thermometers
  • 25 Non-Roll Fittings per pack
PRODUCT OPTIONS
£9.75GBP / Pkg of 1
£11.70GBP (incl VAT)
£183.75GBP / Each
£220.50GBP (incl VAT)
for an additional
£183.75GBP / Each
£220.50GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionLiquid-in-Glass Thermometer Non-Roll Fitting, Purple PVC Plastic, Ring Top
Key Features
  • Can be used for all types of liquid-in-glass thermometers
  • 25 Non-Roll Fittings per pack
More About this Item
Designed prevent glass thermometers from rolling when left on a flat surface. Both styles of anti-roll fittings are made out of durable PVC plastic. Choose from ring top or triangular styles.