This item is now discontinued. Please browse similar items
H-B Instrument B61400-5100 Angled Liquid-in-Glass Thermometer Storage Tray; 14 Slots
Representative image only.
H-B Instrument Mfr # 614005100 – Item # WZ-08007-97
Designed to properly store your glass thermometers at the correct angle
  • Use for all types of liquid-in-glass thermometers
  • Angled to prevent separation of liquid column
  • UV coated
  • 5 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")17 2/5
  • Height (in)1 1/5
  • Overall Length (in)17.4
  • Overall Length (mm)442
  • DescriptionAngled Liquid-in-Glass Thermometer Storage Tray; 14 Slots
  • Warranty5 years
5 Year Warranty
Key Features
  • Use for all types of liquid-in-glass thermometers
  • Angled to prevent separation of liquid column
  • UV coated
  • 5 year warranty
More About this Item
Keep thermometers handy and properly stored while keeping your space organized. Trays are made of 2 mm white polystyrene with an inclined angle to help prevent liquid separation. Each tray has numbered slots. Choose from two sizes - one fits easily in a drawer, the other is suited for table for shelf storage. Sold in sets.