This item is now discontinued. Please browse similar items
H-B Instrument B60303-0100 Easy-Read General Purpose Liquid-In-Glass Thermometer; -10 to 150C, Total Immersion, Environmentally Friendly
Representative image only.
H-B Instrument Mfr # B60303-0100 – Item # WZ-08008-30
Designed to be very easy to read
  • Replace mercury thermometers with the same accuracy without any hazards
  • 100% nontoxic, nonhazardous, biodegradable, and “green"
  • Statement of accuracy included
  • 5 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")7 29/32
  • Min Temperature (° C)-10
  • Max Temperature (° C)150
  • Accuracy±1.0°C; ±1.5°C above 100°C
  • Division (° C)1
  • Thermometer FillBlack Spirit
  • Overall Length (in)7.9
  • Overall Length (mm)200
  • Immersion Depth (mm)200
  • Stem MaterialYellow-Back Glass
  • DescriptionEasy-Read General Purpose Liquid-In-Glass Thermometer; -10 to 150C, Total Immersion, Environmentally Friendly
  • Warranty5 years
5 Year Warranty
Key Features
  • Replace mercury thermometers with the same accuracy without any hazards
  • 100% nontoxic, nonhazardous, biodegradable, and “green"
  • Statement of accuracy included
  • 5 year warranty
More About this Item
These thermometers are ideal for use in laboratories, universities, food/beverage, environmental, wastewater and many other general-purpose applications where the use of mercury or other hazardous liquids is not desired. Each thermometer features a black liquid and yellow back glass which creates a distinct contrast making this thermometer very easy to read. All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. These thermometers come complete with ring top anti-roll fitting, individual serial number, statement of accuracy, recyclable packaging, and are RoHS compliant.