Global Water PL200-H-1 hydrant water pressure logger
Representative image only.
Global Water Mfr # FUA000K – Item # WZ-32922-82
Shaped like a fire hydrant cap to reduce theft or vandalization
  • High-speed sampling mode captures momentary pressure events
  • USB data port and bundled software allow for easy setup, calibration, and data transfer
  • 1 year warranty
PRODUCT OPTIONS
£782.29GBP / Each
£938.75GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Number of Channels1
  • Input TypePressure
  • Max Pressure200 psi
  • Min Pressure0 psi
  • Memory Size81,759 readings
  • Battery Type1 3.6V lithium (included)
  • Height (in)3 7/64
  • Diameter (in)3.9062
  • Descriptionhydrant water pressure logger
  • Warranty1 year
1 Year Warranty
Key Features
  • High-speed sampling mode captures momentary pressure events
  • USB data port and bundled software allow for easy setup, calibration, and data transfer
  • 1 year warranty
More About this Item
Easy-to-use, rugged pressure logger lets you quickly locate distribution pressure problems and gather system modeling data. Massive, nonvolatile memory stores over 81,000 pressure readings. High-speed sampling mode catches events like pressure spikes and water hammer. Flexible datalogging capabilities allow defined interval, high speed, and exception sampling. Bundled software lets user easily export readings via USB data port to a computer. Software additionally provides for easy setup, calibration, and data management.