This item is now discontinued. Please browse similar items
Glas-Col 100E O545 Support for Flexible Fabric Heating Mantle, 3000 Ml
Representative image only.
Glas-Col Mfr # 100E O545 – Item # WZ-36225-86
Safely support flasks and mantles
  • Free up valuable bench space while heating solutions
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Flask Size (mL)3000
  • MaterialSteel
  • DescriptionSupport for Flexible Fabric Heating Mantle, 3000 Ml
  • Warranty1 year
1 Year Warranty
Key Features
  • Free up valuable bench space while heating solutions
  • 1 year warranty
More About this Item
Use these extension supports for attaching hemispherical and spherical mantle to lattice. Each extension is made of a steel ring and heat resistant strap basket.
See our clamps and supports section for support rods and frames.