Gilmont GS-1100 Micrometer Syringe, 0.2 mL; 1/Ea
Representative image only.
Gilmont Item # WZ-07840-00
Dispense small volumes at constant velocity—ideal for titrations and fluid injections
PRODUCT OPTIONS
£274.04GBP / Each
£328.85GBP (incl VAT)
£78.01GBP / Each
£93.61GBP (incl VAT)
for an additional
£78.01GBP / Each
£93.61GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
Cole-Parmer Clean-Wipes, 70% Alcohol/30% DI Water; 100/Canister
£8.50GBP / Pkg of 100
£10.20GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)0.2
  • Graduation Interval (mL)0.0002
  • DescriptionMicrometer Syringe, 0.2 mL; 1/Ea
More About this Item
Turn the handle of this dispenser for precise delivery of small volumes. Dispense fluids with an accuracy of ±0.5% of reading or ±1 scale division, whichever is greater. Fluids only come in contact with the borosilicate glass tip, PTFE syringe barrel, and Viton® O-rings.
Autoclavable
Syringe-style pipettes terminate in a standard luer-lock fitting for easy replacement and interchangeability. Dispensers include one tip.
Optional PTFE O-rings are available separately for applications working with harsh chemicals.