This item is now discontinued. Please browse similar items
Gerber-Method Glass Butyrometer, Milk Test, 11 mL; 1/Pk
Representative image only.
Item # WZ-17425-10
  • measure the percent fat in dairy products
  • made of resistant borosilicate glass
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)11
  • MaterialGlass
  • DescriptionGerber-Method Glass Butyrometer, Milk Test, 11 mL; 1/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • measure the percent fat in dairy products
  • made of resistant borosilicate glass
  • 1 year warranty
More About this Item
Use these butyrometers to measure the percent fat in dairy products. Butyrometers are precision calibrated and feature permanent graduations. Made of resistant borosilicate glass.
Order glass cups for Roeder and Van-Gulik method testing. Stoppers with special acid resistant rubber are available for all butyrometers. Both stopper 17425-15 and stopper pusher 17425-16 are required for milk and Kohler method testing; stoppers 17425-14 and -17 are required for Roeder and Van-Gulik method testing.