This item is now discontinued. Please browse similar items
George Ulanet 301-2 Immersion Heater, Flat, 2.75" Dia. Heated Area, 550 Watts, 230 V
Representative image only.
George Ulanet Mfr # 301-2 – Item # WZ-03049-52
  • 6 Month Product / 12 Month Parts and Labor Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Immersion Depth (in)2
  • Heated Area4.75 in dia
  • Max Temperature (° F)212
  • Max Temperature (° C)100
  • Min TemperatureAmbient;Ambient
  • Plug TypeNo plug
  • Power (VAC)230
  • Power (watts)550
  • DescriptionImmersion Heater, Flat, 2.75\" Dia. Heated Area, 550 Watts, 230 V
  • Warranty6 Month Product / 12 Month Parts and Labor
Key Features
  • 6 Month Product / 12 Month Parts and Labor Warranty
More About this Item
Heaters with Dial Temperature Controller have a sensitivity of ±1°>F (±0.6°>C) in an agitated bath. Features a coiled heating element to concentrate heat at the bottom of the vessel. Heaters include a 6-ft, three-wire cord with plug on 115 VAC models; no plug on 230 VAC models.