This item is now discontinued. Please browse similar items
General Tools LRTH185DL-PROBE 3 ft/1 m extention probe for jumbo LED display temperature/humidity datalogger
Representative image only.
General Tools & Instruments Mfr # LRTH185DLP – Item # WZ-30005-22
Ideal for recording and monitoring environmental parameters at mission-critial applications
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Description3 ft/1 m extention probe for jumbo LED display temperature/humidity datalogger
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
This temperature/humidity datalogger and monitor is made for wall-mounting. With the bright red jumbo display, viewing at intervals is easy and when an audible alarm sounds due to the temperature or humidity parameters moving beyond the set safe-range. This unit is ideal for use in areas where temperature and humidity condition is a critical factor of operation, including labs, electronic manufacturing, and incubators.
You can store up to 30,000 data points with a date and time stamp. Then, download data to a PC via the included USB cable interface for analysis. Powered by AC to DC adapter with a battery backup ensures un-interrupted recording and display of ongoing conditions.
The unit has optional extension probe (30005-22) available which provides an additional 3 feet (1m) of reach. Use this probe to place into closures where you cannot hang the unit itself within.