General Tools DM8252 Digital Manometer, 0/2PSI, High Resolution
Representative image only.
General Tools Mfr # DM8252 – Item # WZ-97008-79
Ideal for laboratory research equipment, computer peripherals, pneumatic controls, HVAC and more
  • Select from one of 5 pressure units of measure: inH2O, psi, mbar, bar, mmH2O
  • Bright backlight
  • Data hold & record features
  • Min/max button
  • Simple fittings for hose connection
  • Auto power off & low battery indicator
  • 1 year warranty
PRODUCT OPTIONS
£286.37GBP / Each
£343.64GBP (incl VAT)
£159.35 - £197.25GBP / Each
£191.22 - £236.70GBP (incl VAT)
for an additional
£159.35GBP / Each
£191.22GBP (incl VAT)
for an additional
£197.25GBP / Each
£236.70GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
Traceable Calibrated Digital Manometer; ±5 psi
£250.86GBP / Each
£301.03GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDgtl Manomtr0/2PSI High Resolution
  • Warranty1 year
1 Year Warranty
Key Features
  • Select from one of 5 pressure units of measure: inH2O, psi, mbar, bar, mmH2O
  • Bright backlight
  • Data hold & record features
  • Min/max button
  • Simple fittings for hose connection
  • Auto power off & low battery indicator
  • 1 year warranty
More About this Item
Displays gauge or differential pressure.