This item is now discontinued. Please browse similar items
General Tools DT8811WP Economical Waterproof Thermocouple Thermometer Type-K
Representative image only.
General Tools Mfr # DT8811WP – Item # WZ-91210-55
Rugged, impact-resistant design that is economical and easy to use
  • Waterproof design — ideal for working in adverse weather conditions
  • Back-lighting button permits reading in dark areas.
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeThermocouple
  • Number Of Probe Channels1
  • Types Of Probes AcceptedType K
  • Min Temperature (° F)-328
  • Max Temperature (° F)2372
  • Min Temperature (° C)-200
  • Max Temperature (° C)1300
  • Resolution0.2°F (0.1°C)
  • IP RatingIP66
  • Low Battery IndicatorYes
  • Display Type1.5\" Display, 3.5-digit LCD
  • Height (in)7
  • Depth (in)1 1/2
  • No. of Probe(s) Included1
  • Battery Life200 hours
  • Battery9v Battery (1)
  • DescriptionEconomical Waterproof Thermocouple Thermometer Type-K
  • Warranty1 year
1 Year Warranty
Key Features
  • Waterproof design — ideal for working in adverse weather conditions
  • Back-lighting button permits reading in dark areas.
  • 1 year warranty
More About this Item
This IP66-rated, ABS plastic thermometer has a 2.5 second sampling rate with an accuracy of 1.8 degrees. Features include min/max, relative temperature, Hold, auto ranging, °F/°C selection, sleep mode, and auto power-off after inactivity to save on battery power. Accepts any type K thermocouple probe with a miniconnector.
What's Included: beaded wire type K probe, hard carrying case, rubber cap, and one 9 V battery.