This item is now discontinued. Please browse similar items
General Tools 800043 Infrared/Thermocouple/RTD Thermometer
Representative image only.
General Tools Mfr # DT2000RTD – Item # WZ-35633-00
A solution for every temperature measurement application
  • One meter uses multiple sensor types
  • Use the temperature sensor best suited to your application
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeMulti-Parameter
  • Number Of Probe Channels1
  • Min Temperature (° F)-4
  • Max Temperature (° F)752
  • Min Temperature (° C)-20
  • Max Temperature (° C)400
  • Accuracy±0.2% of reading +1.0°
  • Min IR Temperature (° F)-328
  • Min IR Temperature (° C)-200
  • Max IR Temperature (° F)2372
  • Max IR Temperature (° C)1300
  • IR Temperature Accuracy±3% of reading +1.0° or ±5.0° (whichever is greater)
  • IR Temperature Resolution (° C)
  • Display Type0.5\" Display
  • Height (in)7 7/8
  • Width (in)2 3/4
  • Depth (in)1 1/4
  • Battery9v Battery (1)
  • DescriptionInfrared/Thermocouple/RTD Thermometer
  • Warranty1 year
1 Year WarrantyConformite EuropeenneLaser Class II
Key Features
  • One meter uses multiple sensor types
  • Use the temperature sensor best suited to your application
  • 1 year warranty
More About this Item
This combination thermometer is a single meter with a solution for any temperature measurement application. For general-purpose applications, choose any type J, K, T, E, or R thermocouple probe; for precise readings, use the 100Ω Pt RTD probe; for noncontact measurements, use the built-in IR thermometer with laser sighting. Thermometer records minimum and maximum temperatures, and features data hold, relative temperature calculations, backlit display, and RS-232 output. Thermometer is easily portable with included carrying case; use on benchtop with convenient tripod back. Meter accepts thermocouples with miniconnectors (sold separately) and 4-pin 100Ω Pt RTD probes.
Cole-Parmer provides a wide offering of Professional Infrared Thermometers with varying features and accuracies depending on your application.
We now offer an FDA-cleared infrared thermometer.
Our other infrared thermometers are not approved for medical usage and are not FDA-approved.
For help in selecting an infrared thermometer please contact the Technical Support team at Cole-Parmer.