This item is now discontinued. Please browse similar items
Gemini 01-14872 Dual Containment Tank for Chemical Feed Systems, 220 Gallons; Blue
Representative image only.
Gemini Mfr # 01-14872 – Item # WZ-06930-30
Exceeds EPA secondary containment requirements (49 CFR)
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Volume (Gal)220
  • Volume (Liters)833
  • Height (in)48
  • Height (cm)121.92
  • Outside Diameter (OD) (in)48
  • Outlet Connections1\" and 2\" NPT(F)
  • DescriptionDual Containment Tank for Chemical Feed Systems, 220 Gallons; Blue
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
These chemical-resistant polypropylene tanks feature heavy-duty wall construction for liquids with specific gravity of 1.5. Tank tops come equipped with an integral pump mounting platform rated to 240 lb (113 kg) — easily mount heavy metering pumps or mixers directly on to the tank lid. Tanks come standard with an 8" vented twist-on lid, 1" or 2" NPT(F) spinwelds, plus a pump suction connection for 3/8" OD tubing, or 1/2" OD tubing when grommet is removed.