This item is now discontinued. Please browse similar items
Fungilab V12010N Small Sample Adapter with Circulation Jacket
Representative image only.
Cole-Parmer Mfr # V12010N – Item # WZ-98965-52
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSmall Sample Adapter with Circulation Jacket
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Expand testing capabilities and accuracy with these viscometer parts and accessories.
The small sample adapter allows the use of smaller volumes of sample. Includes a circulation jacket for connection to an external circulating bath for temperature control.