This item is now discontinued. Please browse similar items
Fuego 6.000.200 Spray Protector (Accessories for Safety Enhanced Laboratory Burners)
Representative image only.
Fuego Mfr # 6.000.200 – Item # WZ-36302-12
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSpray Protector (Accessories for Safety Enhanced Laboratory Burners)
  • Warranty2 Years
Two year warranty
Key Features
  • 2 year warranty
More About this Item
Parts and accessories increase safety and keep your burner in good working order.
Use spray protector with Fuego SCS Safety Enhanced Burners. Shield protects gas flame by minimizing risk from wind currents, drafts and chemical sprays in the laboratory.