This item is now discontinued. Please browse similar items
Fowler 52-580-109 AGD Blackface Dial Indicator/Stand Combo, 0-1"
Representative image only.
Fowler Mfr # 525801090 BOM – Item # WZ-10508-59
High quality multi-function indicator at a reasonable cost
  • Offers direct inch/metric or inch/metric/fraction conversion
  • Easy-to-read dial face
  • Simple to operate
SPECS & DESCRIPTION
Specifications & Description
  • Min range (in)0
  • Max range (in)1
  • Resolution0.001 in
  • DescriptionAGD Blackface Dial Indicator/Stand Combo; 0-1\"
Key Features
  • Offers direct inch/metric or inch/metric/fraction conversion
  • Easy-to-read dial face
  • Simple to operate
More About this Item
These economical battery operated indicators offer absolute/incremental readings on a 2 1/4" digital face. Features include tolerance indicators, floating zero, and carbide tipped point.
Other features of the indicator include: reading: .001", 1" travel, AGD Group 2, and coninuous reading: 0-100. The stand features: 6" x 6" x 2" granite base, positive lock screw, fine adjustment knob for zero setting<, column height is 8", and microscrew fingertip control.