This item is now discontinued. Please browse similar items
Fluke FLK-3561/3502 FC Vibration Sensor Starter Kit with Software, 3 Years
Representative image only.
Fluke Mfr # FLK-3561/3502 FC 3YR – Item # WZ-20001-39
Monitor vibration measurements to predict machine breakdown
  • Remotely measure vibrations in dangerous or hard-to-reach areas for up to three years!
  • Be alerted to changing conditions to prevent machine failure
  • Compact and lightweight
  • IP67 rated—rugged enough for industrial environments
  • Gateway requires Wi-Fi connection and AC power
  • Capture one data point every 90 seconds
  • Includes three year software license
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Vibration Range - Acceleration (g)32
  • Frequency Range10 to 1000 Hz
  • Max Temperature (F)149
  • Max Temperature (C)65
  • Battery Life3 years
  • Battery Type3.6 V lithium
  • DescriptionVibration Sensor/Gateway Starter Kit; 3 Yr License
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Includes
Four vibration sensors, one gateway, FCCM subscription for each sensor, and epoxy.
Key Features
  • Remotely measure vibrations in dangerous or hard-to-reach areas for up to three years!
  • Be alerted to changing conditions to prevent machine failure
  • Compact and lightweight
  • IP67 rated—rugged enough for industrial environments
  • Gateway requires Wi-Fi connection and AC power
  • Capture one data point every 90 seconds
  • Includes three year software license
  • 1 year warranty
More About this Item
Make predictive maintenance fast and easy with the Fluke vibration system consisting of a vibration sensor and gateway. Sensor takes continuous overall vibration measurements of acceleration and velocity for a wide range of machines. Long-term vibration monitoring can help detect imbalance, misalignment, looseness, and bearing faults, saving you catastrophic failures and costly downtime. Install gateway within 65 meters from line-of-sight of sensors. Wi-Fi or mobile router/hotspot connectivity and AC power must be available for gateway to work.
Remotely monitor the conditions using the Fluke Connect Condition Monitoring™ (FCCM) software. Set alarms based on user-defined thresholds or Fluke Overall Vibration Severity (FOVS) scale. FCCM software can send alarms to smart devices or computers within seconds of abnormality. View real-time and historical data to take maintenance action.
Connect up to 20 sensors to one gateway. The number of sensors per gateway is determined by two factors: line of sight and distance from sensors. Select from a starter kit or an expansion kit. Starter kit includes a gateway along with four sensors to get you started on monitoring your equipment. Expansion kit includes four sensors. Each kit is available with 1- or 3-year software licenses.