Fluke 1734/EUS Energy Logger, Advanced with FlukeConnect®, with Flex Probes
Representative image only.
Fluke Mfr # FLUKE-1734/EUS – Item # WZ-20000-93
Discover inefficiencies and optimize your energy bill
  • Automatically capture and log voltage, current, power, power factor, energy and associated values
  • Conserve instrument battery—power instrument directly from the measured circuit
  • Measure all three phases—3 flexible current probes included
  • Perform in-the-field analysis and data checks on the touch screen, graphical display
  • Store more than 20 separate logging sessions
  • CAT IV-600 V/ CAT III-1000 V safety rating
  • Ideal for load studies, energy assessments, and energy surveys
  • 2 year warranty
£4,454.55GBP / Each
£5,345.46GBP (incl VAT)
£246.35GBP / Each
£295.62GBP (incl VAT)
for an additional
£246.35GBP / Each
£295.62GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Max AC Voltage (Voltage)1000
  • Max AC Current (amps)6000
  • Max Frequency Range (Hz)69
  • Frequency Accuracy± (0.1 %)
  • DataloggingNumber of possible logging sessions and logging period depends on user requirements. Typical 10 logging sessions of 8 weeks with 1-minute intervals.
  • MemoryInternal flash memory (not user replaceable)
  • Communication PortsUSB-A, USB-mini, Wi-FI
  • Sample Rate10.24 kHz at 50/60 Hz, synchronized to mains frequency
  • Display4.3-inch active matrix TFT, 480 pixels x 272 pixels, resistive touch panel
  • Battery Type1 3.6V Lithium and 1 1/2 AA (included)
  • DescriptionEnergy Logger, Advanced with FlukeConnect®; EU/US/UK
  • Warranty2 Years
Two year warrantyConformite Europeenne
Includes
voltage test leads, alligator clips (4x), 12 in 1,500A flexible current probe (3x), soft case, Energy Analyze Plus software, magnetic hanging strap, magnetic voltage probes (4x), WiFi/BLE adapter, line cords (EU, US, and UK), color coding set, power supply, and documentation on USB flash drive.
Key Features
  • Automatically capture and log voltage, current, power, power factor, energy and associated values
  • Conserve instrument battery—power instrument directly from the measured circuit
  • Measure all three phases—3 flexible current probes included
  • Perform in-the-field analysis and data checks on the touch screen, graphical display
  • Store more than 20 separate logging sessions
  • CAT IV-600 V/ CAT III-1000 V safety rating
  • Ideal for load studies, energy assessments, and energy surveys
  • 2 year warranty
More About this Item
Find when and where energy in your facility is being consumed—from the service entrance to individual circuits. These energy loggers introduce a new simplicity to discovering sources of electrical energy waste. View data locally on the instrument, through Fluke Connect® mobile app and desktop software, or through your facilities’ Wi-Fi infrastructure. All models are Fluke Connect® compatible, model 20000-93 includes the Wi-Fi/BLE adapter to enable it. Download and analyze every detail of energy consumption with automated reporting. Every session detail is stored so you don't lose measurement trends—automatically.
Available for sale in the United States and Canada only