Flowline XP88-0 Explosion-Proof Ultrasonic Level Transmitter

Representative image only.
Flowline Mfr # XP88-0 – Item # WZ-68873-81
Explosion-proof ultrasonic level transmitter ideal for hazardous applications
  • Compact XP enclosure with viewing window and dual conduit ports
  • Narrow 3'' beam diameter enables flexible installation in applications with limited measurement space
  • Rugged corrosion resistant PVDF transducer and process mount for chemical and wastewater
  • Fail-safe intelligence with diagnostic feedback for easy troubleshooting
  • Display indicates air gap or level height distance in inch or centimeter

£1,382.90GBP / Each
£1,659.48GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Max Depth (ft)24.6
  • Max Depth (m)7.5
  • Min Depth (cm)20
  • Min Depth (in)8
  • Resolution Of Depth Range (in)0.079
  • Resolution Of Depth Range (mm)2
  • Accuracy Of Depth Range±0.2% of max range
  • Dead Band (in)8
  • Signal Output4 to 20 mA
  • Loop Resistance500 Ω max at 24 VDC
  • Display Type6-digit LCD
  • Max Process Temperature (° C)60
  • Min Process Temperature (° F)-4
  • Max Pressure (PSI)30
  • Max Pressure (bar)2
  • Power (VDC)18 to 28
  • DescriptionExplosion-Proof Ultrasonic Level Transmitter

Key Features

  • Compact XP enclosure with viewing window and dual conduit ports
  • Narrow 3'' beam diameter enables flexible installation in applications with limited measurement space
  • Rugged corrosion resistant PVDF transducer and process mount for chemical and wastewater
  • Fail-safe intelligence with diagnostic feedback for easy troubleshooting
  • Display indicates air gap or level height distance in inch or centimeter

More About this Item

EchoSafe™, an explosion proof, two-wire ultrasonic transmitter provides noncontact level measurement up to 10m in hazardous bulk storage, day tank and sump applications, and is well suited for challenging corrosive, slurry or waste media. Media examples include sodium hypochlorite, wastewater and resin. The integral display and push button module enable fast and simple calibration.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat