Flowline LI90-1001 Advanced Controller, 4 analog in/4 relays out
Representative image only.
Flowline Mfr # LI90-1001 – Item # WZ-68587-00
Low-cost controller for industrial applications
  • Graphic tank display for better process and machine visualization
  • Takes less space and is more affordable than seperate PLC and Operator Interfaces
  • First with removable MicroSD™ memory
  • Data logging using built-in MicroSD™ mass data storage
  • Small enough to fit in the palm of your hand, only 3.75” square
£1,023.27GBP / Each
£1,227.92GBP (incl VAT)
£112.50GBP / Each
£135.00GBP (incl VAT)
for an additional
£112.50GBP / Each
£135.00GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionAdvanced Controller, 4 analog in/4 relays out
Key Features
  • Graphic tank display for better process and machine visualization
  • Takes less space and is more affordable than seperate PLC and Operator Interfaces
  • First with removable MicroSD™ memory
  • Data logging using built-in MicroSD™ mass data storage
  • Small enough to fit in the palm of your hand, only 3.75” square
More About this Item
The Commander® advanced controller is an all-in-one solution controller with graphics and removable mass storage for industrial applications starting below the cost of a micro PLC plus operator interface. The commander series is small enough to fit in the palm of your hand, and includes a graphical operator interface and removable mass data storage. One of the first industrial products to include support for the latest standard in removable memory, MicroSD™. Capable of 1 GB of storage, you can use this additional memory to store process data, machine recipes and application files.
From OEM machine control to machine monitoring in a plant environment, the Commander® advanced controller can be instrumental in keeping your automation activities running smoothly. The display is designed with an attractive look so it will fit in with most panel or console designs.