Flowline LC10-1002 Strobe Alert Single Sensor Controller
Representative image only.
Flowline Mfr # LC10-1002 – Item # WZ-43300-43
£243.80GBP / Each
£292.56GBP (incl VAT)
£100.80GBP / Each
£120.96GBP (incl VAT)
for an additional
£100.80GBP / Each
£120.96GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionStrobe Alert Single Sensor Controller
More About this Item
The LC10 AC powered controller is commonly used for the automatic filling or emptying of a tank between two sensors. A single latched relay performs the automatic control with valves or pumps rated up to ½ horsepower. Combine the LC11 controller with Smart Trak and two level switches to complete your compact lift station package. The general purpose, NEMA 4X controller mounts on any sensor or fitting assembly. The optional flash alarm brings immediate attention to process conditions. Package the controller with any of our level switches.
The LC05 compact junction box is offered in both large and small configurations for use with our level or flow switches. The LC05-1001 junction box is commonly used for the integral wiring termination of multiple level switches installed on the Smart Trak fitting. The LC06-1001 junction box is commonly used for the integral wiring termination of a single level or flow switch. The rugged NEMA 4X enclosure includes a 300-degree swivel base for fast and easy conduit alignment.