DescriptionIndustrial TRMS Multimeter with Type K Temperature, Datalogging, and Wireless Connectivity
WarrantyLTD Life Time
Key Features
Built-in thermometer conveniently allows you to take temperature readings without having to carry a separate instrument
Automatically select the optimal measuring range with autoranging feature
Bright, large 6000-count 2.8” digital LCD with bargraph
CAT IV-600 V, CAT III-1000 V safety rating
Rugged IP40 design for enhanced durability
Document measurements with datalogging mode featuring on-board stoarge of 40k measurements
Connect to FLIR IR cameras or mobile devices via Bluetooth
Wirelessly connect to FLIR workflow management applications for streamlining
Limited Lifetime Warranty
More About this Item
These industrial True RMS digital multimeters feature diagnostic measurements including 10 A AC/DC current, 1000 V AC/DC voltage, capacitance, VFD mode, and frequency. Plus, you’ll work more efficiently with advanced features such as diode testing, continuity check, non-contact voltage detection, HOLD function, and a built-in temperature probe for temperature measurement capability. Power your multimeter with alkaline batteries or optional LI-Poly rechargeable battery. For the optional rechargeable battery, order the battery kit separately as part number 20070-01. These multimeters are CAT III 1000 V and CAT IV 600 V rated and can withstand a drop test up to 3 meters. Meter is auto and manual ranging for maximum flexibility during testing. Measurements display numerically and graphically with analog bar graph on the large, backlit LCD. Min/Max and average readings are recorded to capture variations. Choose the DM91 model for added Bluetooth® connectivity and datalogging features.