This item is now discontinued. Please browse similar items
Flir DM62 TRMS Digital Multimeter with Non-Contact Voltage
Representative image only.
Flir Mfr # DM62 – Item # WZ-35725-21
Diagnostic functions provide fast and effective troubleshooting
  • Automatically select the optimal measuring range with autoranging feature
  • Bright, large 6000-count 3.8” digital backlit LCD
  • Durable IP40 design for rugged applications
  • CAT III-600 V, CAT IV-300 V safety rating
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • DC Max (Volts)600
  • DC Voltage Accuracy±0.4%
  • DC Max (amps)10
  • AC Max (Volts)600
  • Ac Voltage Accuracy±1%
  • AC Max (amps)10
  • Resistance Max (KΩ)600
  • Continuity30 to 480 Ω
  • Diode TesterYes
  • True RmsYes
  • Safety RatingCAT IV-300V, CAT III-600V, IP40
  • Power3 AA batteries
  • DescriptionTRMS Digital Multimeter with Non-Contact Voltage
  • WarrantyLTD Life Time
Conformite EuropeenneLimited Lifetime Warranty
Includes
Silicone test leads, integral probe holder, soft carrying case, and batteries.
Key Features
  • Automatically select the optimal measuring range with autoranging feature
  • Bright, large 6000-count 3.8” digital backlit LCD
  • Durable IP40 design for rugged applications
  • CAT III-600 V, CAT IV-300 V safety rating
  • Limited Lifetime Warranty
More About this Item
These True RMS digital multimeters feature diagnostic measurements including 10 A AC/DC current, 600 V AC/DC voltage, and capacitance. Plus, you’ll work more efficiently with advanced features such as diode testing, continuity check, non-contact voltage detection, and min/max/average hold functions.
These multimeters are CAT III 600 V and CAT IV 300 V rated and can withstand a drop test up to 2 meters. Meter is auto and manual ranging for maximum flexibility during testing. Measurements display numerically on the large, backlit LCD with 5 reading per second update rate. Min/Max and average readings are recorded to capture variations.