This item is now discontinued. Please browse similar items
Fisher Accumet AB150 pH Benchtop Meter Kit
Representative image only.
Accumet Mfr # 13636AB150 – Item # WZ-59340-00
Combines basic meter economy with 0.001 pH resolution
  • Plain language prompts for easy setup
  • 500 data points with date-and-time stamp to meet GLP
  • USB and bidirectional RS-232 connectivity
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionpH Benchtop Meter Kit
  • Warranty3 year
3 Year Warranty
Key Features
  • Plain language prompts for easy setup
  • 500 data points with date-and-time stamp to meet GLP
  • USB and bidirectional RS-232 connectivity
  • 3 year warranty
More About this Item
The accumet AB-series meters offer benefits typically found on more expensive meters — a graphic LCD, bidirectional RS-232 and USB connectivity, password protection, and GLP compliant 500 datapoint memory. The large, backlit display lets you easily view measurements with calibration data; plus a graphic electrode status indicator allows you to verify electrode condition at a glance.
The meter accepts up to five-point pH calibration with custom or preset buffer groups in pH mode. It also offers calibration and high/low alarms for system monitoring. ISE mode offers direct measurements in ppm, mg/L, and molar concentration. Meter accepts pH and ISE electrodes with a BNC connector, and two- and four-cell conductivity probes with cell constants equal to 0.1, 1.0, and 10.
Includes TRIS compatible pH/ATC electrode.