Filter Cartridge, Pleated Polyester, 1 micron nominal-rated, 19-1/2" L
Representative image only.
Mfr # HC/90-1 – Item # WZ-29808-26
£145.00GBP / Each
£174.00GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyester
  • Filter MediaPolyester
  • Micron Rating1
  • Filter Length (in)19.5
  • Cartridge Length (in)19.5
  • DescriptionFilter Cartridge, Pleated Polyester, 1 micron nominal-rated, 19-1/2" L
More About this Item
Pleated Polyester Cartridges are general purpose cartridges offering long life and high dirt holding capacity. Polyester cartridges are cleanable and reusable in most applications and are NSF 61 listed.
REVIEWS
Q&A
Do you have questions about this product?
Get answers from real customers and in-house experts with AnswerBox.
1 Question | 1 Answer
Displaying question 1
  • Q:
    For the WU-29808-26 1um filter cartridge. What housing goes with the filter? What is the flow rate?
    Asked on 6/28/2016 by Mike from Gilbert, AZ
    1 answer
    • STAFF EXPERT
      A:
      The flow rate for these cartridges is a maximum of 100GPM. Suggested use is no less than 70GPM. The housings that go with these filters are 00383FN and 00278XE
      Please call our Applications Department directly. We will need to find out more in detail your application in order to recommend the proper filter housing.
      Answered on 8/10/2016 by Dan C from Technical Support
Displaying question 1
Do you have a question about this product? 