Eyela 119170 Vacuum Hose; 16 ft (5 m)

Representative image only.
Eyela Mfr # 119170 – Item # WZ-28625-05
Complete your evaporation system!
  • Vacuum hose
  • 1 year warranty

PRODUCT OPTIONS

£206.22GBP / Each
£247.46GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Compatible WithEyela Industrial Rotary Evaporators
  • Length (in)192
  • Length (cm)50
  • DescriptionVacuum Hose; 16 ft (5 m)
  • Warranty1 year
1 Year Warranty

Key Features

  • Vacuum hose
  • 1 year warranty

More About this Item


Chiller and vacuum pump required for rotary evaporator operation.

Transformer is required for 220 VAC operation. Please contact us for assistance.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat