Extech Sensor Replacement for RD300 Refrigerant Leak Detector
Representative image only.
Extech Mfr # RD300-S – Item # WZ-10112-26
See what you’re doing with handy LED light located right on probe tip
  • Color-coded lights visually indicate leak size
PRODUCT OPTIONS
£75.60GBP / Each
£90.72GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSensor Replacement for RD300 Refrigerant Leak Detector
Includes
leak test bottle, hard carrying case, and one 9 V battery.
Key Features
  • Color-coded lights visually indicate leak size
More About this Item
Detect refrigerant leaks in air conditioning and cooling systems with the Extech RD300 leak detector. A convenient LED work light at the tip of the probe illuminates dimly lit areas, making it easier to identify the source of a leak. Select from three sensitivity levels with multicolored light bars visually indicating the leak size. Mute button toggles the audible alarm on and off. Detector also features a flexible probe, low-battery indication, and auto-shutoff.