Extech PT150 Pressure Transmitter, 150 psi
Representative image only.
Extech Mfr # PT150 – Item # WZ-68602-32
Get fast, realiable measurements
  • Ideal for air or non-corrosive/non-ionized gas
  • 3 year warranty
PRODUCT OPTIONS
£329.32GBP / Each
£395.18GBP (incl VAT)
£159.35 - £197.25GBP / Each
£191.22 - £236.70GBP (incl VAT)
for an additional
£159.35GBP / Each
£191.22GBP (incl VAT)
for an additional
£197.25GBP / Each
£236.70GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Max Pressure (psia)150
  • Wetted MaterialsCorrosion-resistant brass alloy
  • Process Connection1/4\" NPT(M)
  • Electrical Connections3.9 ft ( 1.2 m) cable
  • Output0 to 100mV DC
  • DescriptionPressure Transmitter, 150 psi
  • Warranty3 year
3 Year WarrantyConformite Europeenne
Key Features
  • Ideal for air or non-corrosive/non-ionized gas
  • 3 year warranty
More About this Item
These interchangeable pressure transducers work well with the Extech SDL700 pressure meter/datalogger (model 68602-28).