This item is now discontinued. Please browse similar items
Extech MO280-RK Water Restoration Contractor/Home Inspector Kit
Representative image only.
Extech Mfr # MO280-RK – Item # WZ-86531-12
A complete solution for the Restoration Contractor
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Measurement TypeCombination
  • Moisture Range Reference Scale (Min)0
  • Moisture Range Reference Scale (Max)99
  • Display TypeDigital
  • BatteryBattery (all models include)
  • DescriptionWater Restoration Contractor/Home Inspector Kit
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • 1 year warranty
More About this Item
Kit includes:
Pinless Moisture Meter
Displays %moisture from 0 to 99%
Measurement depth to 0.75" (22mm) below the surface
Wood Moisture Meter
Memory contains 8 wood groups with calibrations for approximately 170 species of wood
Measures wood moisture from 6 to 99.9%
Precision Hygro-Thermometer
Measures Temperature, Humidity, Dew Point, Wet Bulb, Absolute Humidity, GPP
Less than 30 second RH response time with 2%RH accuracy
Humidity/Temperature USB Datalogger
Datalogs 16,000 RH and 16,000 Temperature readings
User-programmable sampling rate and alarm thresholds