This item is now discontinued. Please browse similar items
Extech MO220 Pocket Moisture Meter with Pins and External Probes
Representative image only.
Extech Mfr # MO220 – Item # WZ-86531-51
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Measurement TypePin
  • Moisture Range Reference Scale (Min)6
  • Moisture Range Reference Scale (Max)99.9
  • Temperature CompensationYes, manual or automatic
  • Display TypeDigital
  • BatteryTwo AAA batteries (included)
  • Height (in)8
  • Width (in)2 19/64
  • Length (in)1 45/64
  • DescriptionPocket Moisture Meter with Pins and External Probes
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Monitor moisture in wood and other building materials
  • Designed in a rugged double molded housing construction and is pocket sized for one handed operation
  • Memory contains 9 wood groups with calibrations for appxoimately 170 species of wood
  • Replaceable measurement electrode pins
  • Built-in battery check and measurement verification test
  • Remote temperature probe for automatic temperature compensation
  • Hammer moisture probe for hard materials